missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercaptosuccinic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Mercaptosuccinic acid is used in the preparation of mercaptosuccinic acid diethyl ester by reaction with ethanol. It is also used as a brightening agent in metal plating. Further, it serves as an intermediate for the synthesis of corrosion inhibitors, soil fumigants, active pharmaceutical ingredients and electroplating agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Mercaptosuccinic acid is used in the preparation of mercaptosuccinic acid diethyl ester by reaction with ethanol. It is also used as a brightening agent in metal plating. Further, it serves as an intermediate for the synthesis of corrosion inhibitors, soil fumigants, active pharmaceutical ingredients and electroplating agents.
Solubility
Soluble in water, alcohol and acetone. Slightly soluble in ether. Insoluble in benzene.
Notes
Incompatible with strong oxidizing agents and strong bases.
Mercaptosuccinic acid is used in the preparation of mercaptosuccinic acid diethyl ester by reaction with ethanol. It is also used as a brightening agent in metal plating. Further, it serves as an intermediate for the synthesis of corrosion inhibitors, soil fumigants, active pharmaceutical ingredients and electroplating agents.
Solubility
Soluble in water, alcohol and acetone. Slightly soluble in ether. Insoluble in benzene.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Specifications
Melting Point | 150°C to 155°C |
Quantity | 500 g |
Beilstein | 1099858 |
Merck Index | 14,9343 |
Solubility Information | Soluble in water,alcohol and acetone. Slightly soluble in ether. Insoluble in benzene. |
Formula Weight | 150.15 |
Percent Purity | 98% |
Chemical Name or Material | Mercaptosuccinic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'