Learn More
Mercuric acetate, 98%, Thermo Scientific Chemicals
19.20 € - 893.00 €
Chemical Identifiers
CAS | 1600-27-7 |
---|---|
MDL Number | MFCD00012165 |
InChI Key | BRMYZIKAHFEUFJ-UHFFFAOYSA-L |
Synonym | mercury 2+ ion acetic acid |
PubChem CID | 15337 |
ChEBI | CHEBI:33211 |
IUPAC Name | mercury(2+);diacetate |
SMILES | CC(=O)[O-].CC(=O)[O-].[Hg+2] |
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10327740
|
Thermo Scientific Acros
175020050 |
5 g | Glass Bottle |
19.20 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10741261
|
Thermo Scientific Acros
175021000 |
100 g | Plastic bottle |
69.20 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10741451
|
Thermo Scientific Acros
175025000 |
500 g | Plastic bottle |
228.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10216973
|
Thermo Scientific Acros
175020025 |
2.5 kg | Plastic bottle |
893.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1600-27-7 | |
BRMYZIKAHFEUFJ-UHFFFAOYSA-L | |
15337 | |
mercury(2+);diacetate |
MFCD00012165 | |
mercury 2+ ion acetic acid | |
CHEBI:33211 | |
CC(=O)[O-].CC(=O)[O-].[Hg+2] |
Specifications
179.0°C to 182.0°C | |
White | |
98% | |
(CH3CO2)2Hg | |
02, 96 | |
15, 5940 | |
Solubility in water: 400g/L (20°C). Other solubilities: soluble in alcohol | |
CC(=O)[O-].CC(=O)[O-].[Hg+2] | |
318.68 | |
CHEBI:33211 | |
98% | |
Mercuric acetate, 98% |
1600-27-7 | |
Fine Crystalline Powder | |
C4H6HgO4 | |
MFCD00012165 | |
01,644; 02,264; 03,194; 04,319; 05,424; 06,358; 10,252; 11,315; 12,298; 17,176 | |
mercury 2+ ion acetic acid | |
BRMYZIKAHFEUFJ-UHFFFAOYSA-L | |
mercury(2+);diacetate | |
15337 | |
318.67 | |
Authentic |
Safety and Handling
GHS H Statement:
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Fatal if swallowed.
Fatal in contact with skin.
Very toxic to aquatic life with long lasting effects.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 216-491-1
missing translation for 'rtecsNumber' : AI8575000
missing translation for 'tsca' : TSCA
RUO – Research Use Only