missing translation for 'onlineSavingsMsg'
Learn More

Mercuric Iodide, Powder, Red, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15528034
Click to view available options
Quantity:
125 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 7774-29-0
Color Red
Physical Form Powder
Quantity 125 g
Molecular Formula HgI2
Synonym mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatum
InChI Key YFDLHELOZYVNJE-UHFFFAOYSA-L
SMILES I[Hg]I
IUPAC Name diiodomercury
Molecular Weight (g/mol) 454.401
PubChem CID 24485
ChEBI CHEBI:49659
Formula Weight 454.4
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 6.36kg
Chemical Name or Material Mercuric Iodide
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.