Learn More
Ansell™ Mercury™ 43-113 Series Yellow Aramid Fiber Heavy Weight Gloves Product Code.: 12669721
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
Ansell™ Mercury™ 43-113 Series Yellow Aramid Fiber Heavy Weight Gloves
Product Code. 12669721 Shop All Ansell Products
10
Click to view available options
Size:
10
11
Product Code. 12669721
Brand: Ansell™ 43113100
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
Add to basket
Mercury™ 43-113 Series Yellow Aramid Top of the range gloves for the most exacting hot applications. Also provides fantastic resistance to cuts, tears and abrasion.
  • Soft, comfortable and absorbent
  • Designed for intermittent dry heat contact up to 350°C
  • Knitted construction
  • Cotton Kevlar™ Aramid fibers liner
  • Long knitwrist cuff
TRUSTED_SUSTAINABILITY
Specifications
Back Material Dupont™ Kevlar™ aramid fibers
Cuff Material Dupont™ Kevlar™ aramid fibers
Product Type Heavyweight Gloves
Size 10
Glove Cuff Color Yellow
Glove Cuff Style Long Knitwrist
Lining Cotton
Quantity 72 Pairs
Resistant To Heat, cuts, tears and abrasion resistance
Series 43-113
Product Content Correction
Product Title
Ansell™ Mercury™ 43-113 Series Yellow Aramid Fiber Heavy Weight Gloves >