missing translation for 'onlineSavingsMsg'
Learn More

Mercury, 99.99+%

Product Code. 10712631 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
Packaging:
Plastic bottle
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 10712631

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7439-97-6 | Hg | 200.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Silver
CAS 7439-97-6
Density 13.5400g/mL
Quantity 100 g
Packaging Plastic bottle
Assay Percent Range 99.99+%
Form Heavy Liquid
Molecular Formula Hg
MDL Number MFCD00011035
Merck Index 15, 5966
Synonym hydrargyrum, quicksilver, metallic, liquid silver, mercurio, quecksilber, mercure, quick silver, colloidal, mercury, metallic
InChI Key QSHDDOUJBYECFT-UHFFFAOYSA-N
SMILES [Hg]
IUPAC Name mercury
Molecular Weight (g/mol) 200.59
PubChem CID 23931
ChEBI CHEBI:16170
Formula Weight 200.59
Percent Purity ≥99.990% (MB)
Chemical Name or Material Mercury
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercury
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H330-Fatal if inhaled.
  • H360D-May damage the unborn child.
  • H372-Causes damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.