missing translation for 'onlineSavingsMsg'
Learn More

Mercury(I) chloride, 99.5%, Thermo Scientific Chemicals

For calomel electrodes, in ceramic painting, porcelain painting with gold

51.70 € - 212.00 €

Chemical Identifiers

CAS 10112-91-1
Molecular Formula Cl2Hg2
Molecular Weight (g/mol) 472.084
MDL Number MFCD00011043
InChI Key ZOMNIUBKTOKEHS-UHFFFAOYSA-L
Synonym mercurous chloride, calomel, mercury i chloride, mercury chloride hg2cl2, mercury subchloride, dimercury dichloride, calogreen, calotab, chlorure mercureux, ccris 3803
PubChem CID 24956
ChEBI CHEBI:33050
IUPAC Name chloromercury
SMILES Cl[Hg].Cl[Hg]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11334959
View Documents
Thermo Scientific Alfa Aesar
087240.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11324959
View Documents
Thermo Scientific Alfa Aesar
087240.36
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Mercury(I) chloride is used as calomel electrodes, in ceramic painting and porcelain painting with gold.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10112-91-1
472.084
ZOMNIUBKTOKEHS-UHFFFAOYSA-L
24956
chloromercury
Cl2Hg2
MFCD00011043
mercurous chloride, calomel, mercury i chloride, mercury chloride hg2cl2, mercury subchloride, dimercury dichloride, calogreen, calotab, chlorure mercureux, ccris 3803
CHEBI:33050
Cl[Hg].Cl[Hg]

Specifications

400°C (sublimation)
White
Cl2Hg2
UN2025
mercurous chloride, calomel, mercury i chloride, mercury chloride hg2cl2, mercury subchloride, dimercury dichloride, calogreen, calotab, chlorure mercureux, ccris 3803
ZOMNIUBKTOKEHS-UHFFFAOYSA-L
chloromercury
24956
472.09
Odorless
7.15 g/mL
10112-91-1
Crystalline
MFCD00011043
14,5894
Insoluble in water, alcohol, ether, dilute acids
Cl[Hg].Cl[Hg]
472.084
CHEBI:33050
99.5%
Light and moisture sensitive
Mercury (I) chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H312-H315-H319-H335

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III

missing translation for 'einecsNumber' : 233-307-5

missing translation for 'rtecsNumber' : OV8740000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only