missing translation for 'onlineSavingsMsg'
Learn More

Mercury(I) chloride, ACS reagent, Thermo Scientific Chemicals

Chemical Identifiers

CAS 10112-91-1
Molecular Formula Cl2Hg2
Molecular Weight (g/mol) 472.08
InChI Key ZOMNIUBKTOKEHS-UHFFFAOYSA-L
Synonym mercurous chloride, calomel, mercury i chloride, mercury chloride hg2cl2, mercury subchloride, dimercury dichloride, calogreen, calotab, chlorure mercureux, ccris 3803
PubChem CID 24956
ChEBI CHEBI:33050
IUPAC Name chloromercury
SMILES Cl[Hg].Cl[Hg]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12397193
View Documents Promotion Details
Thermo Scientific Acros
447591000
100 g N/A
N/A
12307203
View Documents Promotion Details
Thermo Scientific Acros
447595000
500 g N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10112-91-1
472.08
mercurous chloride, calomel, mercury i chloride, mercury chloride hg2cl2, mercury subchloride, dimercury dichloride, calogreen, calotab, chlorure mercureux, ccris 3803
CHEBI:33050
Cl[Hg].Cl[Hg]
Cl2Hg2
ZOMNIUBKTOKEHS-UHFFFAOYSA-L
24956
chloromercury

Specifications

400.0°C
Cl2Hg2
15,5962
Solubility in water: practically insoluble. Other solubilities: insoluble in ether and alcohol
Cl[Hg].Cl[Hg]
472.08
CHEBI:33050
ACS Reagent
Mercury(I) chloride
10112-91-1
Hg2Cl2
mercurous chloride, calomel, mercury i chloride, mercury chloride hg2cl2, mercury subchloride, dimercury dichloride, calogreen, calotab, chlorure mercureux, ccris 3803
ZOMNIUBKTOKEHS-UHFFFAOYSA-L
chloromercury
24956
472.08
Plastic bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 233-307-5

Product Identifier
  • Mercury(I) chloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only