missing translation for 'onlineSavingsMsg'
Learn More

Mercury(I) sulfate, 97%, Thermo Scientific Chemicals

26.40 € - 236.00 €

Chemical Identifiers

CAS 7783-36-0
Molecular Weight (g/mol) 497.24
MDL Number MFCD00016138
InChI Key MINVSWONZWKMDC-UHFFFAOYSA-L
Synonym mercurous sulfate, dimercury sulfate, dimercury sulphate, mercury i sulfate, sulfuric acid, dimercury 1+ salt, unii-pi950n9dys, mercurous sulphate, sulfuric acid, dimercury 1++ salt, pi950n9dys
PubChem CID 24545
IUPAC Name mercury(1+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Hg+].[Hg+]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10589423
View Documents Promotion Details
Thermo Scientific Acros
389920050
5 g Plastic bottle
26.40 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10677474
View Documents Promotion Details
Thermo Scientific Acros
389921000
100 g Plastic bottle
236.00 €
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7783-36-0
MFCD00016138
mercurous sulfate, dimercury sulfate, dimercury sulphate, mercury i sulfate, sulfuric acid, dimercury 1+ salt, unii-pi950n9dys, mercurous sulphate, sulfuric acid, dimercury 1++ salt, pi950n9dys
mercury(1+);sulfate
497.24
MINVSWONZWKMDC-UHFFFAOYSA-L
24545
[O-]S(=O)(=O)[O-].[Hg+].[Hg+]

Specifications

7783-36-0
Powder
Hg2O4S
MFCD00016138
mercurous sulfate, dimercury sulfate, dimercury sulphate, mercury i sulfate, sulfuric acid, dimercury 1+ salt, unii-pi950n9dys, mercurous sulphate, sulfuric acid, dimercury 1++ salt, pi950n9dys
MINVSWONZWKMDC-UHFFFAOYSA-L
mercury(1+);sulfate
24545
97%
Beige or White-Yellow
96.0 to 104% (Iodimetry)
Hg2SO4
15, 5965
Solubility in water: 0.06g/100mL (25°C). Other solubilities: soluble in diluted nitric acid
[O-]S(=O)(=O)[O-].[Hg+].[Hg+]
497.24
497.24
Mercury(I) sulfate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Fatal if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 231-993-

Product Identifier
  • Mercury(I) sulfate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H300-Fatal if swallowed.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only