Learn More
Mercury(II) acetate, ACS, 98.0% min, Thermo Scientific Chemicals
44.82 € - 166.83 €
Chemical Identifiers
CAS | 1600-27-7 |
---|---|
Molecular Formula | C4H6HgO4 |
Molecular Weight (g/mol) | 318.68 |
MDL Number | MFCD00012165 |
InChI Key | BRMYZIKAHFEUFJ-UHFFFAOYSA-L |
Synonym | mercury 2+ ion acetic acid |
PubChem CID | 15337 |
ChEBI | CHEBI:33211 |
IUPAC Name | mercury(2+);diacetate |
SMILES | CC(=O)[O-].CC(=O)[O-].[Hg+2] |
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11311619
|
Thermo Scientific Alfa Aesar
012273.18 |
50 g |
44.82 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11301619
|
Thermo Scientific Alfa Aesar
012273.30 |
250 g |
166.83 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mercuric acetate is used as an oxidizing agent in organic synthesis. It is used in oxymercuration of double bonds. Mercuric acetate is used in non-aqueous titration. It is employed in the manufacture of phenyl mercury compounds which have pharmaceutical applications. It removes the acetamidomethyl protecting group from protected thiol, and converts thiocarbonate esters into dithiocarbonates. It promotes the addition of hydroxide and alkoxide across carbon-carbon double bonds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1600-27-7 | |
318.68 | |
BRMYZIKAHFEUFJ-UHFFFAOYSA-L | |
15337 | |
mercury(2+);diacetate |
C4H6HgO4 | |
MFCD00012165 | |
mercury 2+ ion acetic acid | |
CHEBI:33211 | |
CC(=O)[O-].CC(=O)[O-].[Hg+2] |
Specifications
179°C to 182°C | |
Red | |
98.0% min. | |
MFCD00012165 | |
mercury 2+ ion acetic acid | |
BRMYZIKAHFEUFJ-UHFFFAOYSA-L | |
mercury(2+);diacetate | |
15337 | |
318.68 | |
Weak | |
White crystals | |
Mercury(II) acetate |
1600-27-7 | |
Crystalline | |
C4H6HgO4 | |
3563831 | |
Highly soluble in water. Soluble in alcohol and diethyl ether. | |
CC(=O)[O-].CC(=O)[O-].[Hg+2] | |
318.68 | |
CHEBI:33211 | |
ACS | |
Light sensitive | |
3.28 g/cm3 at 20°C |
Safety and Handling
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P312-P314-P330-P361-P363-P501c
H300+H310+H330-H336-H373
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: II
missing translation for 'einecsNumber' : 216-491-1
missing translation for 'rtecsNumber' : AI8575000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Mercury(II) acetate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P361-Take off immediately all contaminated clothing.
Supplemental information
- MIXTURE LIST-Contains : Mercuric acetate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.