missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) bromide, ACS, Thermo Scientific Chemicals

53.16 € - 137.69 €

Chemical Identifiers

CAS 7789-47-1
Molecular Formula Br2Hg
Molecular Weight (g/mol) 360.4
MDL Number MFCD00011040
InChI Key NGYIMTKLQULBOO-UHFFFAOYSA-L
Synonym mercuric bromide, mercury dibromide, mercury ii bromide, hgbr2, mercury bromide hgbr2, mercuric dibromide, mercury ii bromide 1:2, hsdb 829, hg2, mercury 2 bromide
PubChem CID 24612
ChEBI CHEBI:49639
IUPAC Name dibromomercury
SMILES Br[Hg]Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11450111
SDS View Documents
Thermo Scientific Alfa Aesar
044816.14
25 g
53.16 €
25g
Please to purchase this item. Need a web account? Register with us today!
11460111
SDS View Documents
Thermo Scientific Alfa Aesar
044816.22
100 g
137.69 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Mercury(II) bromide is used as a reagent in the Koenigs-Knorr reaction, which forms glycoside linkages on carbohydrates. It is also used to test for the presence of arsenic, as recommended by the Pharmacopoeia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-47-1
360.4
NGYIMTKLQULBOO-UHFFFAOYSA-L
24612
dibromomercury
Br2Hg
MFCD00011040
mercuric bromide, mercury dibromide, mercury ii bromide, hgbr2, mercury bromide hgbr2, mercuric dibromide, mercury ii bromide 1:2, hsdb 829, hg2, mercury 2 bromide
CHEBI:49639
Br[Hg]Br

Specifications

322°C to 325°C (sublimation)
7789-47-1
Powder
MFCD00011040
Soluble in hot alcohol,methanol,HCl,HBr
Br[Hg]Br
360.4
CHEBI:49639
ACS
Light sensitive
6.05 g/cm3 at 20°C
236°C
White
Br2Hg
mercuric bromide, mercury dibromide, mercury ii bromide, hgbr2, mercury bromide hgbr2, mercuric dibromide, mercury ii bromide 1:2, hsdb 829, hg2, mercury 2 bromide
NGYIMTKLQULBOO-UHFFFAOYSA-L
dibromomercury
24612
360.4
Odorless
1mm Hg at 137°C
Mercury(II) bromide, For ACS analysis
Videos
Safety and Handling

Safety and Handling

P260-P262-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P310-P314-P330-P361-P363-P501c

H300+H310+H330-H373

missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: II

missing translation for 'einecsNumber' : 232-169-3

missing translation for 'rtecsNumber' : OV7415000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Mercuric bromide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.