missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) chloride, 98+%, Thermo Scientific Chemicals

Product Code. 11428077
Click to view available options
Quantity:
50 g
250 g
1000 g

The main application of mercuric chloride is as a catalyst for the conversion of acetylene to vinyl chloride, the precursor to polyvinylchloride. It is also employed in photography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The main application of mercuric chloride is as a catalyst for the conversion of acetylene to vinyl chloride, the precursor to polyvinylchloride. It is also employed in photography, disinfectants, wood preservatives

Solubility
Soluble in diethyl ether. Insoluble in water.

Notes
Store at room temperature. Reactive with oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7487-94-7
Molecular Formula Cl2Hg
Molecular Weight (g/mol) 271.49
MDL Number MFCD00011041
InChI Key LWJROJCJINYWOX-UHFFFAOYSA-L
Synonym mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2
PubChem CID 24085
ChEBI CHEBI:31823
IUPAC Name dichloromercury
SMILES [Cl-].[Cl-].[Hg++]

Specifications

Boiling Point 302°C
Melting Point 280°C to 282°C
Quantity 1000 g
UN Number UN1624
Merck Index 14,5876
Solubility Information Soluble in diethyl ether. Insoluble in water.
Formula Weight 271.5
Percent Purity ≥98%
Odor Odorless
Refractive Index 1.859
Density 5.4
Chemical Name or Material Mercury(II) chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercury(II) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H341-Suspected of causing genetic defects.
  • H361f-Suspected of damaging fertility.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Mercuric chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.