Learn More
Mercury(II) chloride, 99.5+%, ACS reagent, Thermo Scientific Chemicals
56.38 € - 681.39 €
Chemical Identifiers
CAS | 7487-94-7 |
---|---|
Molecular Formula | Cl2Hg |
Molecular Weight (g/mol) | 271.49 |
MDL Number | MFCD00011041 |
InChI Key | LWJROJCJINYWOX-UHFFFAOYSA-L |
Synonym | mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2 |
PubChem CID | 24085 |
ChEBI | CHEBI:31823 |
IUPAC Name | dichloromercury |
SMILES | [Cl-].[Cl-].[Hg++] |
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10687381
|
Thermo Scientific Acros
423931000 |
100 g | Plastic bottle |
56.38 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10219800
|
Thermo Scientific Acros
423935000 |
500 g | Plastic bottle |
175.88 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10013783
|
Thermo Scientific Acros
423930025 |
2.5 kg | Plastic bottle |
681.39 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7487-94-7 | |
271.49 | |
LWJROJCJINYWOX-UHFFFAOYSA-L | |
24085 | |
dichloromercury |
Cl2Hg | |
MFCD00011041 | |
mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2 | |
CHEBI:31823 | |
[Cl-].[Cl-].[Hg++] |
Specifications
7487-94-7 | |
Crystalline Powder and/or Crystals | |
Cl2Hg | |
MFCD00011041 | |
15, 5943 | |
LWJROJCJINYWOX-UHFFFAOYSA-L | |
dichloromercury | |
24085 | |
271.5 | |
ACS Reagent | |
Mercury(II) chloride |
White | |
99.5+% | |
HgCl2 | |
01,652; 05,427; 06,359; 09,291; 11,467; 12,304; 13,175; 15,200 | |
mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2 | |
[Cl-].[Cl-].[Hg++] | |
271.49 | |
CHEBI:31823 | |
≥99.5% | |
5.44 |
Safety and Handling
GHS P Statement
Fatal if swallowed.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.
Suspected of damaging fertility.
Causes damage to organs through prolonged or repeated exposure.
GHS P Statement
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 231-299-8
missing translation for 'rtecsNumber' : OV9100000
missing translation for 'tsca' : TSCA
- Mercury(II) chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H300-Fatal if swallowed.
- H314-Causes severe skin burns and eye damage.
- H341-Suspected of causing genetic defects.
- H361f-Suspected of damaging fertility.
- H372-Causes damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.