Learn More
Mercury(II) chloride, ACS, 99.5% min, Thermo Scientific Chemicals
50.15 € - 372.86 €
Chemical Identifiers
CAS | 7487-94-7 |
---|---|
Molecular Formula | Cl2Hg |
Molecular Weight (g/mol) | 271.49 |
MDL Number | MFCD00011041 |
InChI Key | LWJROJCJINYWOX-UHFFFAOYSA-L |
Synonym | mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2 |
PubChem CID | 24085 |
ChEBI | CHEBI:31823 |
IUPAC Name | dichloromercury |
SMILES | [Cl-].[Cl-].[Hg++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11341619
|
Thermo Scientific Alfa Aesar
012274.14 |
25 g |
50.15 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11331619
|
Thermo Scientific Alfa Aesar
012274.22 |
100 g |
88.24 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11321619
|
Thermo Scientific Alfa Aesar
012274.36 |
500 g |
372.86 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mercury(II) chloride is used as a catalyst in making plastics, depolarizer in batteries and as a reagent in organic synthesis and analytical chemistry. It is also used to prepare an amalgam with metals such as aluminum, which finds application as reducing agent in organic synthesis. It also finds application in photography as a photographic intensifier. Further, it serves as a preservative and used in the treatment of syphilis. It is also used to remove dithiane groups bonded to a carbonyl in an umpolung reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7487-94-7 | |
271.49 | |
LWJROJCJINYWOX-UHFFFAOYSA-L | |
24085 | |
dichloromercury |
Cl2Hg | |
MFCD00011041 | |
mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2 | |
CHEBI:31823 | |
[Cl-].[Cl-].[Hg++] |
Specifications
302°C | |
7487-94-7 | |
Powder | |
Cl2Hg | |
mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2 | |
LWJROJCJINYWOX-UHFFFAOYSA-L | |
dichloromercury | |
24085 | |
271.5 | |
Odorless | |
White powder | |
5.4 g/cm3 at 20°C |
280°C to 282°C | |
White | |
99.5% min. | |
MFCD00011041 | |
Soluble in boiling water (solubility in cold water is increased by HCl or alkali chlorides),alcohol,glycerol,methanol,acetone,and ethyl acetateSoluble in water,alcohol,acetone and ethyl acetate. Slightly soluble in benzene,carbon disulfide and pyridine. | |
[Cl-].[Cl-].[Hg++] | |
271.49 | |
CHEBI:31823 | |
ACS | |
1.859 | |
0.0001 hPa | |
Mercury(II) chloride |
Safety and Handling
Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
P201-P202-P260-P262-P264b-P270-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H300-H310-H314-H335-H341-H361f-H372
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: II
missing translation for 'einecsNumber' : 231-299-8
missing translation for 'rtecsNumber' : OV9100000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only