missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury(Ii) Chloride, puriss. p.a. ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 500 g |
Linear Formula | HgCl2 |
UN Number | 1624 |
Formula Weight | 271.5g/mol |
Percent Purity | ≥99.5% (calc. to the dried substance) |
Grade | ACS/puriss. p.a. |
Vapor Pressure | 1.3mmHg (236°C) |
Chemical Name or Material | Mercury(Ii) Chloride, Reag. ISO |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'