missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury(II) iodide, ACS, 99.0% min (Assay-dried basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
As an analytical reagent for the preparation of Nessler's Reagent. It is also used as an oxidizing agent for the synthesis of quinoxaline. Polycrystalline films of mercury (II) iodide, also play an important role in X-ray imaging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | 350°C (sublimation) |
Melting Point | 259°C |
Color | White |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | 99.0% min. |
Solubility Information | Insoluble in water. Soluble in boiling alcohol,ether,acetone,ethyl acetate,carbon disulfide,alkali iodides. |
Formula Weight | 454.4 |
Grade | ACS |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'