Learn More
Mercury(II) iodide, ACS, 99.0% min (Assay-dried basis), Thermo Scientific Chemicals
For Use As An Analytical Reagent For the Preparation of Nessler'S Reagent.
89.40 € - 335.00 €
Chemical Identifiers
CAS | 7774-29-0 |
---|---|
Molecular Formula | HgI2 |
Molecular Weight (g/mol) | 454.401 |
MDL Number | MFCD00011044 |
InChI Key | YFDLHELOZYVNJE-UHFFFAOYSA-L |
Synonym | mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatum |
PubChem CID | 24485 |
ChEBI | CHEBI:49659 |
IUPAC Name | diiodomercury |
SMILES | I[Hg]I |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11381639
|
Thermo Scientific Alfa Aesar
012289.22 |
100 g |
89.40 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11371639
|
Thermo Scientific Alfa Aesar
012289.36 |
500 g |
335.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
As an analytical reagent for the preparation of Nessler's Reagent. It is also used as an oxidizing agent for the synthesis of quinoxaline. Polycrystalline films of mercury (II) iodide, also play an important role in X-ray imaging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7774-29-0 | |
454.401 | |
YFDLHELOZYVNJE-UHFFFAOYSA-L | |
24485 | |
diiodomercury |
HgI2 | |
MFCD00011044 | |
mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatum | |
CHEBI:49659 | |
I[Hg]I |
Specifications
350°C (sublimation) | |
7774-29-0 | |
Powder | |
HgI2 | |
mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatum | |
YFDLHELOZYVNJE-UHFFFAOYSA-L | |
diiodomercury | |
24485 | |
454.4 | |
Odorless | |
Red powder or beads | |
Mercury(II) iodide, Assay-dried basis |
259°C | |
White | |
99.0% min. | |
MFCD00011044 | |
Insoluble in water. Soluble in boiling alcohol,ether,acetone,ethyl acetate,carbon disulfide,alkali iodides. | |
I[Hg]I | |
454.401 | |
CHEBI:49659 | |
ACS | |
Light sensitive | |
6.28 g/cm3 at 20°C |
Safety and Handling
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
H300+H310+H330-H373
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: II
missing translation for 'einecsNumber' : 231-873-8
missing translation for 'rtecsNumber' : OW5250000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only