missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) nitrate hydrate, ACS, 98.0% min, Thermo Scientific Chemicals

Product Code. 11376479
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Mercury(II) nitrate is used as an oxidizing agent involved in organic synthesis. Its reaction with acetone yielded an organometallic compound containing mercury. In addition, it serves as a nitrification agent and analytical reagents used in the laboratory. It is involved in the preparation of mercury fulminate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-34-8
Molecular Formula H2HgN2O7
Molecular Weight (g/mol) 342.62
MDL Number MFCD00149736
InChI Key KVICROHOONHSRH-UHFFFAOYSA-N
Synonym mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2
PubChem CID 3084029
IUPAC Name mercury(2+) hydrate dinitrate
SMILES O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 79°C
Color White
Physical Form Crystalline
Quantity 250 g
Linear Formula Hg(NO3)2·xH2O (x=1 to 2)
UN Number UN1625
Merck Index 14,5880
Solubility Information Soluble in water.
Formula Weight 324.60 (Anhydrous)
Percent Purity ≤98.0%
Grade ACS
Sensitivity Hygroscopic
Density 4.39 g/mL
Chemical Name or Material Mercury(II) nitrate hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'