missing translation for 'onlineSavingsMsg'
Learn More
Mercury(II) nitrate monohydrate, ACS reagent, Thermo Scientific Chemicals Product Code.: 10321261

Mercury(II) nitrate monohydrate, ACS reagent, Thermo Scientific Chemicals

Product Code. 10321261
100 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10321261

Brand: Thermo Scientific Acros 423941000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-34-8
Molecular Formula H2HgN2O7
Molecular Weight (g/mol) 342.62
MDL Number MFCD00149736
InChI Key KVICROHOONHSRH-UHFFFAOYSA-N
Synonym mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2
PubChem CID 3084029
IUPAC Name mercury(2+) hydrate dinitrate
SMILES O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 77.0°C to 79.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 100 g
Linear Formula Hg(NO3)2·H2O
Fieser 03,197; 10,254; 11,317
Merck Index 14, 5880
Solubility Information Solubility in water: slightly soluble
Formula Weight 342.6
Percent Purity ≥98.0%
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 4.3
Density 4.3000g/mL
Chemical Name or Material Mercury(II) nitrate monohydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468827 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number423945000,423941000
Lot NumberA0464367 Certificate TypeCertificate of Analysis Date10/07/2024 Catalog Number423945000,423941000
Lot NumberA0455830 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number423945000,423941000
Lot NumberA0396906 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number423945000,423941000
Lot NumberA0439368 Certificate TypeCertificate of Analysis Date23/02/2022 Catalog Number
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercury(II) nitrate monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mercury(II) nitrate monohydrate, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.