missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury(II) Nitrate Solution, 0.005M, Honeywell™
Click to view available options
Quantity:
1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
CAS | 7783-34-8 |
Quantity | 1 L |
Molecular Formula | H2HgN2O7 |
Linear Formula | Hg(NO3)2 |
MDL Number | MFCD00149736 |
Synonym | mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2 |
InChI Key | KVICROHOONHSRH-UHFFFAOYSA-N |
SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Hg+2] |
IUPAC Name | mercury(2+);dinitrate;hydrate |
Molecular Weight (g/mol) | 342.62 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'