missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) Nitrate Solution, 0.05M, Honeywell™

Product Code. 15624160
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Volumetric, 0.05 M Hg(NO3)2 (0.1N), for the determination of chloride

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Specifications

CAS 7783-34-8
Quantity 1 L
Molecular Formula H2HgN2O7
Linear Formula Hg(NO3)2
MDL Number MFCD00149736
UN Number UN3082
Synonym mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2
InChI Key KVICROHOONHSRH-UHFFFAOYSA-N
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Hg+2]
IUPAC Name mercury(2+);dinitrate;hydrate
Molecular Weight (g/mol) 342.62
PubChem CID 3084029
Formula Weight 324.6g/mol
Concentration or Composition (by Analyte or Components) 0.05M
Chemical Name or Material Mercury(II) nitrate solution
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'