Learn More
Mercury(II) oxide, red, ACS, 99.0% min, Thermo Scientific Chemicals
Description
It is used in marine and porcelain paints, dry batteries, chemical reagents, and topical antiseptics. It acts as a significant part in the production of mercury. It is utilitarian in the deduction of a mercury based superconductor. It is likewise employed, as a photo-oxidant for the photochemical oxidation of primary aromatic amines to the corresponding azoaromatic compounds. It is also an essential material for the preparation of cathodes in mercury batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Orange-Red |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 99.0% min. |
Formula Weight | 216.59 |
Grade | ACS |
Odor | Odorless |
Appearance | Dark orange powder |
Chemical Name or Material | Mercury(II) oxide, red |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.