missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) oxide, yellow, ACS, 99.0% min, Thermo Scientific Chemicals

Product Code. 11313328
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Mercury(II) oxide production of mercury. It is also used as a material for cathodes for mercury batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21908-53-2
Molecular Formula HgO
Molecular Weight (g/mol) 216.59
MDL Number MFCD00011045
InChI Key UKWHYYKOEPRTIC-UHFFFAOYSA-N
Synonym mercuric oxide, mercury ii oxide, mercury oxide, mercuric oxide, red, red mercuric oxide, mercuric oxide, yellow, santar, red precipitate, mercury monoxide
IUPAC Name oxomercury
SMILES O=[Hg]

Specifications

Melting Point 500°C (decomposition)
Color Orange
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.0% min.
Solubility Information Insoluble in water. Insoluble in alcohol,ether,acetone,ammonia.
Formula Weight 216.59
Grade ACS
Appearance Dark orange
Density 11.14
Chemical Name or Material Mercury(II) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'