missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) oxide, yellow, Reagent Grade, Thermo Scientific Chemicals

Product Code. 11351619
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as a chemical intermediate for mercury salts, organic mercury compounds, and chlorine monoxide, antiseptic in pharmaceuticals, component of dry cell batteries, pigment and glass modifier, fungicide, preservative in cosmetics, analytical reagent, formerly used in antifouling paints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21908-53-2
Molecular Formula HgO
Molecular Weight (g/mol) 216.59
MDL Number MFCD00011045
InChI Key UKWHYYKOEPRTIC-UHFFFAOYSA-N
Synonym mercuric oxide, mercury ii oxide, mercury oxide, mercuric oxide, red, red mercuric oxide, mercuric oxide, yellow, santar, red precipitate, mercury monoxide
IUPAC Name oxomercury
SMILES O=[Hg]

Specifications

Melting Point 500°C (decomposition)
Color Yellow
Physical Form Powder
Quantity 100 g
UN Number UN1641
Merck Index 14,5883
Solubility Information Insoluble in water alcohol,ether,acetone,alkali,ammonia.Soluble in most acids.
Formula Weight 216.59
Grade Reagent
Odor Odorless
Density 11.14 g/mL
Chemical Name or Material Mercury(II) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.