missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) perchlorate trihydrate, Reagent Grade, Thermo Scientific™

Product Code. 11321738
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73491-34-6
Molecular Formula Cl2H6HgO11
Molecular Weight (g/mol) 453.529
MDL Number MFCD00149735
InChI Key DFJWPGPUWHLJIJ-UHFFFAOYSA-L
Synonym mercury ii perchlorate trihydrate, mercury perchlorate-water 1/2/3, perchloric acid,mercury 2+ salt, trihydrate 9ci
PubChem CID 53471339
IUPAC Name mercury(2+);diperchlorate;trihydrate
SMILES O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Hg+2]

Specifications

Color White
Physical Form Crystalline
Quantity 100 g
Linear Formula Hg(ClO4)2·3H2O
UN Number UN3087
Solubility Information Soluble in water. Insoluble in benzene and hexane.
Formula Weight 453.54 (399.49 Anhydrous)
Grade Reagent
Chemical Name or Material Mercury(II) perchlorate trihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.