Learn More
Mercury(II) selenide, 99.999% (metals basis), Thermo Scientific Chemicals
80.40 € - 290.00 €
Chemical Identifiers
CAS | 20601-83-6 |
---|---|
Molecular Formula | HgSe |
Molecular Weight (g/mol) | 279.56 |
MDL Number | MFCD00049743 |
InChI Key | YQMLDSWXEQOSPP-UHFFFAOYSA-N |
Synonym | mercury selenide, mercury ii selenide, mercury-selenium complex, mercury monoselenide, mercury 2+ selenide, mercury ii selenide trace metals basis 1g |
PubChem CID | 88609 |
IUPAC Name | selanylidenemercury |
SMILES | [Se]=[Hg] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11317567
|
Thermo Scientific Alfa Aesar
012832.03 |
1 g |
80.40 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11327567
|
Thermo Scientific Alfa Aesar
012832.06 |
5 g |
290.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mercury(II) selenide is used in semiconductors, infrared detectors, solar cells, transistors and amplifiers. It is also used as filters in some steel plants in order to remove mercury from exhaust gases. Further, it is used as an ohmic contact to wide-gap II-VI semiconductors such as zinc selenide or zinc oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
20601-83-6 | |
279.56 | |
YQMLDSWXEQOSPP-UHFFFAOYSA-N | |
88609 | |
[Se]=[Hg] |
HgSe | |
MFCD00049743 | |
mercury selenide, mercury ii selenide, mercury-selenium complex, mercury monoselenide, mercury 2+ selenide, mercury ii selenide trace metals basis 1g | |
selanylidenemercury |
Specifications
(sublimation) | |
Gray | |
HgSe | |
UN2025 | |
Insoluble in water. | |
[Se]=[Hg] | |
279.56 | |
279.55 | |
Odorless | |
(metals basis) |
20601-83-6 | |
Powder | |
MFCD00049743 | |
mercury selenide, mercury ii selenide, mercury-selenium complex, mercury monoselenide, mercury 2+ selenide, mercury ii selenide trace metals basis 1g | |
YQMLDSWXEQOSPP-UHFFFAOYSA-N | |
selanylidenemercury | |
88609 | |
99.999% | |
8.21 g/mL | |
Mercury(II) selenide |
Safety and Handling
GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c
H301-H330-H360Df-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.
missing translation for 'einecsNumber' : 243-910-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only