missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) silver iodide, Thermo Scientific™

Product Code. 11323718
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-03-4
Molecular Formula Ag2HgI4
Molecular Weight (g/mol) 923.94
MDL Number MFCD00046160
InChI Key YNTJRFXPYRRRKS-UHFFFAOYSA-J

Specifications

Color Yellow
Physical Form Powder
Quantity 5 g
UN Number UN2025
Formula Weight 923.95
Sensitivity Light sensitive
Chemical Name or Material Mercury(II) silver iodide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'