Learn More
Mercury(II) sulfate, 98+%, Thermo Scientific Chemicals
53.97 € - 583.91 €
Chemical Identifiers
CAS | 7783-35-9 |
---|---|
Molecular Formula | HgO4S |
Molecular Weight (g/mol) | 296.65 |
MDL Number | MFCD00011047 |
InChI Key | DOBUSJIVSSJEDA-UHFFFAOYSA-L |
Synonym | mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate |
PubChem CID | 24544 |
SMILES | [Hg++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11417807
|
Thermo Scientific Alfa Aesar
A16330.18 |
50 g |
53.97 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11427807
|
Thermo Scientific Alfa Aesar
A16330.30 |
250 g |
196.98 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11407807
|
Thermo Scientific Alfa Aesar
A16330.0B |
1000 g |
583.91 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in medicine as a catalyst and in extracting gold and silver.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in medicine as a catalyst and in extracting gold and silver.
Solubility
It is soluble in hot H2SO4, NaCl solution. insoluble in alcohol, acetone, ammonia
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Light sensitive. Moisture sensitive. Air sensitive. Keep away from Strong oxidizing agents.
Chemical Identifiers
7783-35-9 | |
296.65 | |
DOBUSJIVSSJEDA-UHFFFAOYSA-L | |
24544 |
HgO4S | |
MFCD00011047 | |
mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate | |
[Hg++].[O-]S([O-])(=O)=O |
Specifications
(decomposition) | |
HgO4S | |
MFCD00011047 | |
14,5887 | |
It is soluble in hot H2SO4,NaCl solution. insoluble in alcohol,acetone,ammonia | |
[Hg++].[O-]S([O-])(=O)=O | |
296.65 | |
296.65 | |
Moisture sensitive | |
Mercury(II) sulfate |
7783-35-9 | |
HgSO4 | |
UN1645 | |
mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate | |
DOBUSJIVSSJEDA-UHFFFAOYSA-L | |
mercury(2+);sulfate | |
24544 | |
≥98% | |
6.47 |
Safety and Handling
GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P312-P314-P330-P361-P363-P501c
H300+H310+H330-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY SULFATES
missing translation for 'einecsNumber' : 231-992-5
missing translation for 'rtecsNumber' : OX0500000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Mercury(II) sulphate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
Supplemental information
- MIXTURE LIST-Contains : Sulphuric acid, mercury(2+) salt
RUO – Research Use Only