missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) sulfate, 99+%, Thermo Scientific Chemicals

Product Code. 11904391
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-35-9
Molecular Formula HgO4S
Molecular Weight (g/mol) 296.65
MDL Number MFCD00011047
InChI Key DOBUSJIVSSJEDA-UHFFFAOYSA-L
Synonym mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
PubChem CID 24544
SMILES [Hg++].[O-]S([O-])(=O)=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 99% min
Linear Formula HgSO4
Fieser 01,658; 15,2
Merck Index 15,5955
Solubility Information Solubility in water: reacts. Other solubilities: soluble in hcl, hot dil. h2so4 and sol. of nacl
IUPAC Name mercury(2+) sulfate
Formula Weight 296.64
Percent Purity ≥99%
Packaging Plastic bottle
Specific Gravity 6.47
Density 6.4700g/mL
Chemical Name or Material Mercury(II) sulfate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercury(II) sulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'