missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) sulfate, Honeywell Fluka™

Product Code. 15687280
Click to view available options
Quantity:
10 g
50 g
100 g
250 g
500 g
3 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥99.0% (precipitation titration)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-35-9
Molecular Formula HgO4S
Molecular Weight (g/mol) 296.65
MDL Number MFCD00011047
InChI Key DOBUSJIVSSJEDA-UHFFFAOYSA-L
Synonym mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
PubChem CID 24544
IUPAC Name mercury(2+);sulfate
SMILES [Hg++].[O-]S([O-])(=O)=O

Specifications

Quantity 500 g
Linear Formula HgSO4
UN Number UN1645
Formula Weight 296.65g/mol
Percent Purity ≥99% (precipitation titration)
Chemical Name or Material Mercury(II) sulfate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'