missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) sulfide, black, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

180.00 € - 650.00 €

Chemical Identifiers

CAS 1344-48-5
Molecular Formula HgS
Molecular Weight (g/mol) 232.65
MDL Number MFCD00011046
InChI Key LGZXYFMMLRYXLK-UHFFFAOYSA-N
Synonym mercuric sulfide, mercury ii sulfide, vermilion, mercuric sulfide red, almaden, cinnabar, ethiops mineral, mercury sulfide, red cinnabar, mercury sulphide
PubChem CID 62402
IUPAC Name sulfanylidenemercury
SMILES [S--].[Hg++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11372088
View Documents
Thermo Scientific Alfa Aesar
012992.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11382088
View Documents
Thermo Scientific Alfa Aesar
012992.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Mercury(II) sulfide is used in paint, sealing wax and preservatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1344-48-5
232.65
LGZXYFMMLRYXLK-UHFFFAOYSA-N
62402
[S--].[Hg++]
HgS
MFCD00011046
mercuric sulfide, mercury ii sulfide, vermilion, mercuric sulfide red, almaden, cinnabar, ethiops mineral, mercury sulfide, red cinnabar, mercury sulphide
sulfanylidenemercury

Specifications

583.5°C (sublimation)
Black
HgS
14,5888
Soluble in water (0.00001g/L).
[S--].[Hg++]
232.65
232.65
Puratronic™
7.70 g/mL
Mercury(II) sulfide
1344-48-5
Powder
MFCD00011046
mercuric sulfide, mercury ii sulfide, vermilion, mercuric sulfide red, almaden, cinnabar, ethiops mineral, mercury sulfide, red cinnabar, mercury sulphide
LGZXYFMMLRYXLK-UHFFFAOYSA-N
sulfanylidenemercury
62402
99.999%
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c

H300+H310+H330-H373

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II

missing translation for 'einecsNumber' : 215-696-3

missing translation for 'rtecsNumber' : OX0720000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.