missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) sulfide, red, Thermo Scientific Chemicals

151.00 € - 805.01 €

Chemical Identifiers

CAS 1344-48-5
Molecular Formula HgS
Molecular Weight (g/mol) 232.65
MDL Number MFCD00011046
InChI Key LGZXYFMMLRYXLK-UHFFFAOYSA-N
Synonym mercuric sulfide, mercury ii sulfide, vermilion, mercuric sulfide red, almaden, cinnabar, ethiops mineral, mercury sulfide, red cinnabar, mercury sulphide
PubChem CID 62402
SMILES [S--].[Hg++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11336858
View Documents
Thermo Scientific Alfa Aesar
013482.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11346858
View Documents
Thermo Scientific Alfa Aesar
013482.36
500 g
805.01 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Mercury(II) sulfide, red is used chiefly as a pigment in paints, rubber and plastics. It is also used as a semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1344-48-5
232.65
LGZXYFMMLRYXLK-UHFFFAOYSA-N
62402
HgS
MFCD00011046
mercuric sulfide, mercury ii sulfide, vermilion, mercuric sulfide red, almaden, cinnabar, ethiops mineral, mercury sulfide, red cinnabar, mercury sulphide
[S--].[Hg++]

Specifications

583.5° (sublimation)
Red
HgS
mercuric sulfide, mercury ii sulfide, vermilion, mercuric sulfide red, almaden, cinnabar, ethiops mineral, mercury sulfide, red cinnabar, mercury sulphide
LGZXYFMMLRYXLK-UHFFFAOYSA-N
sulfanylidenemercury
62402
Odorless
7.70 g/cm3
1344-48-5
Powder
MFCD00011046
Insoluble in water.
[S--].[Hg++]
232.65
232.65
Red powder
Mercury(II) sulfide
Videos
Safety and Handling

Safety and Handling

P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c

H300+H310+H330-H373

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II

missing translation for 'einecsNumber' : 215-696-3

missing translation for 'rtecsNumber' : OX0720000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.