missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) telluride, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11364177
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Mercury cadmium telluride (MCT)is the mostim- portantmaterial currently in use for the manufacture of high performance infrared (IR) detectors. Mercury telluride and mercury telluride-cadmium telluride are semiconductors which have been investigated as intrinsic infrared detecting materials. Complete coverage of the low-loss fiber windows by mercury telluride quantum dot are used in colloidal nanocrystals for telecommunications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12068-90-5
Molecular Formula HgTe
Molecular Weight (g/mol) 328.19
MDL Number MFCD00016133
InChI Key VCEXCCILEWFFBG-UHFFFAOYSA-N
Synonym mercury telluride, mercury ii telluride, mercuric telluride, quecksilbertellurid, mercurous telluride, mercury ii telluride trace metals basis 5g
PubChem CID 82914
IUPAC Name tellanylidenemercury
SMILES [Te]=[Hg]

Specifications

Melting Point 673°C
Color Black to Gray
Physical Form Fused Lump
Quantity 2 g
UN Number UN2025
Solubility Information Insoluble in water.
Formula Weight 328.19
Percent Purity 99.999%
Odor Odorless
Density 8.63 g/mL
Assay (metals basis)
Chemical Name or Material Mercury(II) telluride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'