missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) thiocyanate, Thermo Scientific Chemicals

74.83 € - 327.63 €

Chemical Identifiers

CAS 592-85-8
Molecular Formula C4HgK2N4S4
Molecular Weight (g/mol) 511.10
MDL Number MFCD00011039
InChI Key HWXOCCNUUYRBKP-UHFFFAOYSA-J
IUPAC Name mercury(2+) dipotassium tetrakis(cyanosulfanide)
SMILES [K+].[K+].[Hg++].[S-]C#N.[S-]C#N.[S-]C#N.[S-]C#N
View More Specs

Alert:

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.


Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11394277
SDS View Documents
Thermo Scientific Alfa Aesar
037106.18
50 g
74.83 €
50g
Please to purchase this item. Need a web account? Register with us today!
11384277
SDS View Documents
Thermo Scientific Alfa Aesar
037106.30
250 g
327.63 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Mercury(II) thiocyanate is used as a precursor to potassium tris(thiocyanato)mercurate(II) and cesium tris(thiocyanato)mercurate(II). It is also used in the determination of chloride ions in water by UV-visible spectroscopy. Further, it acts as a catalyst for the addition of thiocyanic acid to alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

592-85-8
511.10
HWXOCCNUUYRBKP-UHFFFAOYSA-J
[K+].[K+].[Hg++].[S-]C#N.[S-]C#N.[S-]C#N.[S-]C#N
C4HgK2N4S4
MFCD00011039
mercury(2+) dipotassium tetrakis(cyanosulfanide)

Specifications

∽165°C (decomposition)
White to Yellow
C4HgK2N4S4
3561344
HWXOCCNUUYRBKP-UHFFFAOYSA-J
mercury(2+) dipotassium tetrakis(cyanosulfanide)
316.75
Hygroscopic
3.718 g/cm3 at 20°C
592-85-8
Powder
MFCD00011039
Soluble in dilute hydrochloric acid,potassium cyanide and ammonia. Slightly soluble in alcohol and ether. Insoluble in water.
[K+].[K+].[Hg++].[S-]C#N.[S-]C#N.[S-]C#N.[S-]C#N
511.10
Odorless
White to pale yellow powder
Mercury(II) thiocyanate
Videos
Safety and Handling

Safety and Handling

P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c

H300+H310+H330-H373

missing translation for 'dotInformation' : DOT Class: 6.1; Packing Group: II

missing translation for 'einecsNumber' : 209-773-0

missing translation for 'rtecsNumber' : XL1550000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' : Hygroscopic

Recommended Storage : Ambient temperatures

Product Identifier
  • Mercury(II) thiocyanate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

Supplemental information
  • MIXTURE LIST-Contains : Mercuric thiocyanate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.