missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals MES, 1.0M buffer soln., pH 5.0 Product Code.: 15484439

Thermo Scientific Chemicals MES, 1.0M buffer soln., pH 5.0

Product Code. 15484439
250 mL
Click to view available options
Quantity:
250 mL
500 mL
This item is not returnable. View return policy

Product Code. 15484439

Brand: Thermo Scientific Chemicals J61960.AK

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MES is used as a buffering agent in biology and biochemistry. It is utilized for resolving very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES is used as a buffering agent in biology and biochemistry. It is utilized for resolving very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.248
MDL Number MFCD00283848
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
PubChem CID 16218417
IUPAC Name 2-morpholin-4-ylethanesulfonic acid;hydrate
SMILES C1COCCN1CCS(=O)(=O)O.O

Specifications

Concentration 1.0 M
pH 5.0
Physical Form Liquid
Quantity 250 mL
Solubility Information Miscible with water.
Chemical Name or Material MES buffer soln.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM31L539 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberJ61960.AP,J61960.AK
Lot NumberM11L577 Certificate TypeCertificate of Analysis Date31/12/2024 Catalog NumberJ61960.AP,J61960.AK
Lot NumberX20K536 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog NumberJ61960.AP,J61960.AK
Lot NumberR24K500 Certificate TypeCertificate of Analysis Date28/06/2024 Catalog NumberJ61960.AP,J61960.AK
Lot NumberQ04K535 Certificate TypeCertificate of Analysis Date07/05/2024 Catalog NumberJ61960.AP,J61960.AK

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals MES, 1.0M buffer soln., pH 5.0 >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.