missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MES monohydrate, 99+%, for biochemistry

145.73 € - 975.86 €

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 195.23
MDL Number MFCD00149409
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
Synonym 2-(N-Morpholino)ethanesulfonic acid monohydrate
PubChem CID 16218417
IUPAC Name 2-morpholin-4-ylethanesulfonic acid;hydrate
SMILES C1COCCN1CCS(=O)(=O)O.O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10517241
SDS View Documents
Thermo Scientific Chemicals
327761000
100 g Plastic bottle
145.73 €
100g
Please to purchase this item. Need a web account? Register with us today!
10781804
SDS View Documents
Thermo Scientific Chemicals
327765000
500 g Plastic bottle
395.97 €
500g
Please to purchase this item. Need a web account? Register with us today!
10297403
SDS View Documents
Thermo Scientific Chemicals
327760025
2.5 kg Plastic bottle
975.86 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

Product Identifier
  • MES monohydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals MES monohydrate, 99+%, for biochemistry

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.