missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MES-SDS running buffer (20X)

105.53 € - 200.00 €

Specifications

Concentration 20X
Physical Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15434569
View Documents
Thermo Scientific Chemicals
J62138.AP
500 mL
105.53 €
500mL
Please to purchase this item. Need a web account? Register with us today!
15444569
View Documents
Thermo Scientific Chemicals
J62138.K2
1 L
200.00 €
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MES-SDS running buffer (20X) is used for high resolution of proteins on neutral pH. It finds application in the separation of small to medium-sized proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES-SDS running buffer (20X) is used for high resolution of proteins on neutral pH. It finds application in the separation of small to medium-sized proteins.

Solubility
Miscible with water.

Notes
This buffer contains: 1M MES, 1M Tris base,2% SDS and 20mM EDTA at pH 7.3. Store at room temperature. Keep away from light. Incompatible with strong oxidizing agent and acids.
Specifications

Specifications

20X
Liquid
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

Product Identifier
  • MES-SDS running buffer (20X)

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Sodium lauryl sulfate, EDTA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only