missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals MES sodium salt, 99%
55.58 € - 564.81 €
Chemical Identifiers
CAS | 71119-23-8 |
---|---|
Molecular Formula | C6H12NNaO4S |
Molecular Weight (g/mol) | 217.215 |
MDL Number | MFCD00065473 |
InChI Key | IRHWMYKYLWNHTL-UHFFFAOYSA-M |
PubChem CID | 23673676 |
ChEBI | CHEBI:62955 |
IUPAC Name | sodium;2-morpholin-4-ylethanesulfonate |
SMILES | C1COCCN1CCS(=O)(=O)[O-].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10585714
|
Thermo Scientific Chemicals
397350250 |
25 g | Plastic bottle |
55.58 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10776465
|
Thermo Scientific Chemicals
397351000 |
100 g | Plastic bottle |
150.75 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10421284
|
Thermo Scientific Chemicals
397355000 |
500 g | Plastic bottle |
564.81 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
71119-23-8 | |
217.215 | |
IRHWMYKYLWNHTL-UHFFFAOYSA-M | |
CHEBI:62955 | |
C1COCCN1CCS(=O)(=O)[O-].[Na+] |
C6H12NNaO4S | |
MFCD00065473 | |
23673676 | |
sodium;2-morpholin-4-ylethanesulfonate |
Specifications
0.03 max. (10% in water) 1 cm cell vs water at 280nm, 0.05 max. (10% in water) 1 cm cell vs water at 260nm | |
White | |
99% | |
C6H12NNaO4S | |
Solubility in water: 0.5g/L. | |
C1COCCN1CCS(=O)(=O)[O-].[Na+] | |
217.215 | |
CHEBI:62955 | |
≥98.5% (after ion exchange) |
71119-23-8 | |
Powder | |
Authentic | |
MFCD00065473 | |
IRHWMYKYLWNHTL-UHFFFAOYSA-M | |
sodium;2-morpholin-4-ylethanesulfonate | |
23673676 | |
217.22 | |
MES sodium salt |
Safety and Handling
Product Identifier
- MES sodium salt
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title