Learn More
Manutan Mesh locker, 20 compartment without doors, 1370mm x 1515mm x 305mm
Product Code. 12804259
Encompass_Preferred
Product Code. 12804259
Brand: Manutan A330048
Made from strong steel, perfect where ventilation and high visual security are required. Manufactured from 25x25x2.5mm steel welded mesh on 8mm round bar frame. All units fitted with 150mm integral legs that can be securely fixed to the floor. Finish: bright zinc plated with a clear coat of lacquer. 12, 16 and 20 Compartment units are supplied with individual doors per compartment. 40 Compartment units are supplied with double doors. Includes 3-point locking with hasp and staple for use with padlocks.
  • 20 compartments without doors
  • Dimensions (H x W x D): 1370mm x 1515mm x 305mm
  • Compartment dimensions (H x W x D): 300mm x 300mm x 300mm