Learn More
Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
76.38 € - 278.39 €
Chemical Identifiers
CAS | 141-79-7 |
---|---|
Molecular Formula | C6H10O |
Molecular Weight (g/mol) | 98.14 |
InChI Key | SHOJXDKTYKFBRD-UHFFFAOYSA-N |
Synonym | mesityl oxide, 4-methyl-3-penten-2-one, 3-penten-2-one, 4-methyl, isopropylideneacetone, isobutenyl methyl ketone, methyl isobutenyl ketone, mesityloxid, mesityloxyde, 3-isohexen-2-one, isopropylidene acetone |
PubChem CID | 8858 |
IUPAC Name | 4-methylpent-3-en-2-one |
SMILES | CC(=CC(=O)C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10356860
|
Thermo Scientific Acros
125600010 |
1 L | Glass Bottle |
76.38 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10366860
|
Thermo Scientific Acros
125600025 |
2.5 L | Glass Bottle |
97.18 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11904821
|
Thermo Scientific Acros
125600100 |
10 L | Plastic drum |
278.39 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
141-79-7 | |
98.14 | |
mesityl oxide, 4-methyl-3-penten-2-one, 3-penten-2-one, 4-methyl, isopropylideneacetone, isobutenyl methyl ketone, methyl isobutenyl ketone, mesityloxid, mesityloxyde, 3-isohexen-2-one, isopropylidene acetone | |
4-methylpent-3-en-2-one |
C6H10O | |
SHOJXDKTYKFBRD-UHFFFAOYSA-N | |
8858 | |
CC(=CC(=O)C)C |
Specifications
141-79-7 | |
0.8540g/mL | |
28°C | |
90% alpha-Isomer (GC) | |
(CH3)2C=CHCOCH3 | |
01,736 | |
0.854 | |
Solubility in water: 28g/L (20°C). Other solubilities: miscible with most common organic solvents | |
CC(=CC(=O)C)C | |
98.14 | |
0.64 mPa.s (20°C) | |
99% | |
Mesityl oxide |
-53°C | |
129°C | |
Authentic | |
C6H10O | |
1.4420 to 1.446 | |
15,5978 | |
mesityl oxide, 4-methyl-3-penten-2-one, 3-penten-2-one, 4-methyl, isopropylideneacetone, isobutenyl methyl ketone, methyl isobutenyl ketone, mesityloxid, mesityloxyde, 3-isohexen-2-one, isopropylidene acetone | |
SHOJXDKTYKFBRD-UHFFFAOYSA-N | |
4-methylpent-3-en-2-one | |
8858 | |
98.14 | |
Liquid |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 205-502-5
Recommended Storage : Product may darken during storage
- Mesityl oxide
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.