missing translation for 'onlineSavingsMsg'
Learn More

Mesitylene, 99%, Extra Pure

Product Code. 10625752 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
This item is not returnable. View return policy

Product Code. 10625752

missing translation for 'mfr': Thermo Scientific Acros 125582500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-67-8 | C9H12 | 120.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-67-8
Molecular Formula C9H12
Molecular Weight (g/mol) 120.19
MDL Number MFCD00008538
InChI Key AUHZEENZYGFFBQ-UHFFFAOYSA-N
Synonym mesitylene, sym-trimethylbenzene, benzene, 1,3,5-trimethyl, fleet-x, trimethylbenzol, 3,5-dimethyltoluene, s-trimethylbenzene, hsdb 92, 2,4,6-trimethylbenzene, 1,3,5-trimethyl-benzene
PubChem CID 7947
ChEBI CHEBI:34833
IUPAC Name 1,3,5-trimethylbenzene
SMILES CC1=CC(=CC(=C1)C)C

Specifications

Melting Point -45.0°C
Density 0.8675g/mL
Boiling Point 163.0°C to 166.0°C
Flash Point 44°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H3(CH3)3
Refractive Index 1.4980 to 1.5000
Quantity 250 mL
Beilstein 05, 406
Merck Index 15, 5977
Specific Gravity 0.8675
Solubility Information Solubility in water: 2.9g/L (20°C). Other solubilities: miscible with alcohol,benzene and ether
Viscosity 0.834 mm2/s (20°C)
Formula Weight 120.19
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Mesitylene
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mesitylene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.