missing translation for 'onlineSavingsMsg'
Learn More

meso-2,3-Dimercaptosuccinic acid, 98%

Product Code. 10553894 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10553894

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 304-55-2 | C4H6O4S2 | 182.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304-55-2
Molecular Formula C4H6O4S2
Molecular Weight (g/mol) 182.21
MDL Number MFCD00064799
InChI Key ACTRVOBWPAIOHC-XIXRPRMCSA-N
Synonym succimer, meso-2,3-dimercaptosuccinic acid, meso-dimercaptosuccinic acid, dmsa, dim-sa, 2r,3s-rel-2,3-dimercaptosuccinic acid, succimero, succimerum, chemet, dimercaptosuccinic acid
PubChem CID 2724354
ChEBI CHEBI:63623
IUPAC Name (2S,3R)-2,3-bis(sulfanyl)butanedioic acid
SMILES C(C(C(=O)O)S)(C(=O)O)S

Specifications

Melting Point 196°C to 198°C
Color White
Loss on Drying 1% max. (1 g, 105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HO2CCH(SH)CH(SH)CO2H
Quantity 100 g
Merck Index 15,8995
Formula Weight 182.21
Percent Purity 98%
Physical Form Powder
Chemical Name or Material meso-2, 3-Dimercaptosuccinic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.