missing translation for 'onlineSavingsMsg'
Learn More
Learn More
meso-Hydrobenzoin, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
meso- Hydrobenzoin has been used in the preparation of trans-methyl meso- hydrobenzoin phosphite.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
meso- Hydrobenzoin has been used in the preparation of trans-methyl meso- hydrobenzoin phosphite.
Solubility
Soluble in hot alcohol or chloroform.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
meso- Hydrobenzoin has been used in the preparation of trans-methyl meso- hydrobenzoin phosphite.
Solubility
Soluble in hot alcohol or chloroform.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Specifications
Specifications
Melting Point | 134°C to 136°C |
Quantity | 5 g |
Beilstein | 2050814 |
Merck Index | 14,4777 |
Solubility Information | Soluble in hot alcohol or chloroform. |
Formula Weight | 214.27 |
Percent Purity | 99% |
Chemical Name or Material | meso-Hydrobenzoin |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction