missing translation for 'onlineSavingsMsg'
Learn More

meta-Phosphoric Acid, ≥33.5%, Honeywell™ Fluka™, BioXtra

Product Code. 15617960
Click to view available options
Quantity:
1 L
Packaging:
Poly bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

BioXtra,≥33.5%

  • Stringent testing protocols may include trace metal analysis and specialized suitability testing, such as hybridoma or mouse embryo
  • Includes application-specific formulations, such as tablets and prepared solutions.
TRUSTED_SUSTAINABILITY

Specifications

pH 2 (at 33.00g/L)
Color Colorless
CAS 7647-01-0
Melting Point 200°C
Boiling Point 600°C (1013hPa)
Density 1.2g/mL at 25°C
Packaging Poly bottle
Vapor Pressure 3.23psi (21.1°C), 7.93psi (37.7°C)
Quantity 1 L
Molecular Formula ClH
Physical Form Solid
MDL Number MFCD00011324 MFCD00792839
UN Number UN1789
Beilstein 1098214
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Solubility Information Soluble
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46g/mol
Percent Purity 35.0 to 39.0%
Chemical Name or Material Hydrochloric acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.