missing translation for 'onlineSavingsMsg'
Learn More

Meta-Phosphoric Acid, ACS Reagent, Honeywell Fluka™

Product Code. 15632440
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
6 x 500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 37267-86-0
Molecular Formula HO3P
Molecular Weight (g/mol) 79.979
MDL Number MFCD00011351
InChI Key UEZVMMHDMIWARA-UHFFFAOYSA-N
Synonym metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, hydrogen phosphonate
PubChem CID 3084658
SMILES OP(=O)=O

Specifications

Concentration 33.5 to 36.5%
Quantity 5 g
Linear Formula (HPO3)n
UN Number 3260
Grade ACS Reagent
Chemical Name or Material Meta-Phosphoric Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.