missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Metabolic Activity Dead Cell Apoptosis Kit with C12 Resazurin, Annexin V APC, and SYTOX™ Green, for flow cytometry

Product Code. 10532703
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10532703

Brand: Molecular Probes™ V35114

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vybrant™ Apoptosis Assay Kit #10 - allophycocyanin annexin V/C12-resazurin/SYTOX™ Green - 50 assays

This flow cytometry product provides a three-color fluorescence assay that distinguishes live, apoptotic, and late apoptotic cells from one another. These populations can easily be distinguished using a flow cytometer that has both 488 nm and 633 nm excitation sources (an argon-ion laser and a HeNe laser) and the following reagents Annexin V to detect phosphatidylserine, C12 resazurin for cell metabolism, and SYTOX™ Green nucleic acid stain for compromised membranes.

View a selection guide for all apoptosis assays for flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 kit
Format Tube, Slide
Product Type Dead Cell Apoptosis Kit
Content And Storage Contains 1 vial of annexin V, APC conjugate (250 μL), 1 vial of SYTOX™ green stain (100 μL), 1 vial of C12-resazurin (40 μg), and 1 vial of DMSO (1.5 mL).

Store in refrigerator (2–8°C) and protect from light.
Flow Cytometer Laser Lines 633/635, 488
Excitation/Emission C12-Resazurin: 571⁄585, SYTOX™ green: 503⁄524, APC: 650⁄660
No. of Reactions 50 reactions
Shipping Condition Wet Ice
Product Line SYTOX
For Use With (Application) Flow Cytometry
Conjugate APC, SYTOX Green, C12-resazurin
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.