missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aroclor 1016 in Methyl-tert.butylether 1000μg/mL, Fisher Chemical™
Shop All Fisher Chemical Products

Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Packaging | Ampoule |
| CAS | 12674-11-2 |
| Molecular Formula | C12H7Cl3 |
| Concentration | 1000 μg/mL |
| InChI Key | RIBGNAJQTOXRDK-UHFFFAOYSA-N |
| Type | Organic Single-component Standard Solution |
| SMILES | C1=CC(=CC(=C1)Cl)C2=CC(=CC(=C2)Cl)Cl |
| IUPAC Name | 1,3-dichloro-5-(3-chlorophenyl)benzene |
| Solvent or Matrix | MTBE |
| Molecular Weight (g/mol) | 257.538 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction