missing translation for 'onlineSavingsMsg'
Learn More

Aroclor 1016 in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12996003
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Concentration 100μg/mL
CAS 12674-11-2
Packaging Ampoule
Quantity 1 mL
Type Organic Single-component Standard Solution
Molecular Formula C12H7Cl3
InChI Key RIBGNAJQTOXRDK-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)Cl)C2=CC(=CC(=C2)Cl)Cl
IUPAC Name 1,3-dichloro-5-(3-chlorophenyl)benzene
Molecular Weight (g/mol) 257.538
PubChem CID 38037
Solvent or Matrix Methanol
Chemical Name or Material Aroclor 1016
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.