missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Metacycline hydrochloride, 98%, sec. Standard

Product Code. 10308790 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
Packaging:
Glass Bottle
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 10308790

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3963-95-9
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C22H22N2O8·HCl
MDL Number MFCD04972012
Quantity 100 mg
Merck Index 15, 6015
Synonym methacycline hydrochloride, methacycline hcl, adriamicina, ciclobiotic, germiciclin, rondomycin, metadomus, metacycline hydrochloride, globociclina, metilenbiotic
InChI Key VXPSARQTYDZXAO-CCHMMTNSSA-N
SMILES CN(C)C1C2C(C3C(=C)C4=C(C(=CC=C4)O)C(=C3C(=O)C2(C(=C(C1=O)C(=O)N)O)O)O)O.Cl
IUPAC Name (4S,4aR,5S,5aR,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methylidene-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
Molecular Weight (g/mol) 478.9
PubChem CID 54685047
Formula Weight 478.9
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Metacycline hydrochloride, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.