Learn More
Sampling Systems™ Metal Detectable Pallet Knives and Stirrers
BIOPHARMA
Metal Detectable Pallet Knives and Stirrers are ideal for use with powders, pastes, gels and granules.
Brand:  Sampling Systems™ M1016B-200
Product Code. 17544716
  • 21.55 € / Pack of 4
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The detectable stirrer and pallet knives are made from a special chemically resistant grade of PP.
  • The robust design means they can be used for years and years or used as a single use product straight from the bag.
  • The whole range is manufactured in cleanroom conditions and packed in a class 7 (10,000) cleanroom.
  • This ensures the purity and cleanliness of the product.
  • Simply unwrap and use.
  • Our ergonomic handle means they are easy to use.
  • Many customers use these as stirrers.
  • Made from high quality PP.
  • Gives long lasting performance.
  • Conforms to the latest FDA and EU food requirements.
  • 200 mm blade is a standard size.
Specifications
Specifications
Polypropylene
Blue
200 mm