missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Hose Barb Pump Connector

Fits flexible tubing. Grant Instruments™ Hose Barb Pump Connector is for use with Grant Refrigerated Baths and Heating Circulators

Brand:  Grant Instruments™ M-HB9

 View more versions of this product

Product Code. 15112605

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Metal pump inlet/outlet connector
  • Hose barb stlyle fits flexible tubing
  • Temperature range -40 to 120°C
Specifications

Specifications

Fits Flexible Tubing 9mm I.D
Pump Connector
9 mm
-40°C to +120°C
Metal
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.